The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_110601.1 | 2 - 3 | 383 | 86 | |
NR_110602.1 | 2 - 3 | 345 | 86 | ||
XM_017025364.1 | XP_016880853.1 | 2 - 3 | 357 | 86 | |
XM_017025365.1 | XP_016880854.1 | 2 - 3 | 309 | 86 | |
GenBank mRNA | AK297156.1 | - | 2 - 3 | 301 | 86 |
AK316405.1 | - | 2 - 3 | 84 | ||
BC035662.1 | - | 2 - 3 | 344 | 86 |