Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYTH1
Entrez Gene ID
9267
Gene Name
cytohesin 1
Gene Aliases
B2-1, CYTOHESIN-1, D17S811E, PSCD1, SEC7
Chromosome Location
Chr.17: 78674047 - 78782342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292018.1 NP_001278947.1 11 - 12 1406 92
NM_001292019.1 NP_001278948.1 11 - 12 972 92
NM_004762.3 NP_004753.1 11 - 12 1009 92
NM_017456.3 NP_059430.2 10 - 11 1006 92
XM_011525475.2 XP_011523777.1 10 - 11 868 92
XM_011525476.2 XP_011523778.1 10 - 11 868 92
XM_011525477.1 XP_011523779.1 9 - 10 787 92
XM_011525478.1 XP_011523780.1 10 - 11 1328 92
XM_017025336.1 XP_016880825.1 10 - 11 1076 92
XM_017025337.1 XP_016880826.1 10 - 11 961 92
XM_017025338.1 XP_016880827.1 10 - 11 1392 92
GenBank mRNA AK293894.1 - 11 - 12 1406 92
AK296914.1 - 8 - 9 534 92
AK307917.1 - 11 - 12 938 92
AK316277.1 - 11 - 12 953 92
BC038385.1 - 11 - 12 930 92
BC050452.1 - 11 - 12 919 92
M85169.1 - 11 - 12 959 92

View Full Product Details