The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001292018.1 | NP_001278947.1 | 2 - 3 | 624 | 101 |
NM_001292019.1 | NP_001278948.1 | 2 - 3 | 190 | 101 | |
NM_004762.3 | NP_004753.1 | 2 - 3 | 227 | 101 | |
NM_017456.3 | NP_059430.2 | 2 - 3 | 227 | 101 | |
XM_011525475.2 | XP_011523777.1 | 1 - 2 | 86 | 101 | |
XM_011525476.2 | XP_011523778.1 | 1 - 2 | 86 | 101 | |
XM_011525478.1 | XP_011523780.1 | 2 - 3 | 613 | 101 | |
XM_017025336.1 | XP_016880825.1 | 2 - 3 | 297 | 101 | |
XM_017025337.1 | XP_016880826.1 | 2 - 3 | 182 | 101 | |
XM_017025338.1 | XP_016880827.1 | 2 - 3 | 613 | 101 | |
GenBank mRNA | AK293894.1 | - | 2 - 3 | 624 | 101 |
AK316075.1 | - | 2 - 3 | 183 | 101 | |
BC038385.1 | - | 2 - 3 | 148 | 101 | |
BC050452.1 | - | 2 - 3 | 137 | 101 | |
BX339155.2 | - | 2 - 3 | 207 | 101 | |
DA125333.1 | - | 2 - 3 | 190 | 101 | |
HY049953.1 | - | 2 - 3 | 227 | 101 | |
M85169.1 | - | 2 - 3 | 177 | 101 |