The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135920.2 | NP_001129392.2 | 4 - 5 | 630 | 92 |
NM_001206557.1 | NP_001193486.1 | 4 - 5 | 630 | 92 | |
NM_020829.3 | NP_065880.2 | 4 - 5 | 630 | 92 | |
XM_005251523.3 | XP_005251580.1 | 4 - 5 | 719 | 92 | |
XM_017014934.1 | XP_016870423.1 | 4 - 5 | 286 | 92 | |
XM_017014935.1 | XP_016870424.1 | 2 - 3 | 1187 | 92 | |
XM_017014936.1 | XP_016870425.1 | 2 - 3 | 126 | 92 | |
GenBank mRNA | AB205401.1 | - | 3 - 4 | 224 | 92 |
AK124394.1 | - | 1 - 2 | 1158 | 92 | |
AL136875.1 | - | 4 - 5 | 422 | 92 | |
BC136616.1 | - | 3 - 4 | 224 | 92 | |
BC144296.1 | - | 3 - 4 | 224 | 92 |