The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001257359.1 | NP_001244288.1 | 5 - 6 | 965 | 78 |
NM_174920.3 | NP_777580.1 | 5 - 6 | 965 | 78 | |
XM_011524490.2 | XP_011522792.1 | 2 - 3 | 151 | 78 | |
XM_017024322.1 | XP_016879811.1 | 4 - 5 | 1349 | 78 | |
XM_017024323.1 | XP_016879812.1 | 6 - 7 | 1132 | 78 | |
XM_017024324.1 | XP_016879813.1 | 5 - 6 | 1820 | 78 | |
GenBank mRNA | AK094209.1 | - | 5 - 6 | 896 | 78 |
AK095809.1 | - | 2 - 3 | 266 | 78 | |
AK225767.1 | - | 5 - 6 | 896 | 78 | |
AK291482.1 | - | 5 - 6 | 892 | 78 | |
AY148482.1 | - | 5 - 6 | 886 | 78 | |
BC029872.1 | - | 1 - 2 | 77 | 78 | |
BC047081.1 | - | 4 - 5 | 619 | 78 | |
BC136485.1 | - | 5 - 6 | 878 | 78 | |
BC141822.1 | - | 5 - 6 | 905 | 78 | |
BC142728.1 | - | 5 - 6 | 862 | 78 |