Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD14
Entrez Gene ID
201191
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
-
Chromosome Location
Chr.17: 50111309 - 50129882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257359.1 NP_001244288.1 7 - 8 1202 83
XM_011524490.2 XP_011522792.1 4 - 5 388 83
XM_017024322.1 XP_016879811.1 6 - 7 1586 83
XM_017024323.1 XP_016879812.1 8 - 9 1369 83
GenBank mRNA AK094209.1 - 7 - 8 1133 83
AK095809.1 - 4 - 5 503 83
AK225767.1 - 7 - 8 1133 83
AK291482.1 - 7 - 8 1129 83
BC029872.1 - 3 - 4 314 83
BC047081.1 - 6 - 7 856 83
BC136485.1 - 7 - 8 1115 83
BC141822.1 - 7 - 8 1142 83
BC142728.1 - 7 - 8 1099 83

View Full Product Details