The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006868.3 | NP_006859.2 | 1 - 2 | 215 | 67 |
GenBank mRNA | AF183421.1 | - | 1 - 2 | 168 | 67 |
AF234995.1 | - | 1 - 2 | 214 | 67 | |
AF498957.1 | - | 1 - 2 | 37 | 67 | |
AK223505.1 | - | 1 - 2 | 79 | 67 | |
AK314809.1 | - | 1 - 2 | 173 | 67 | |
BC001148.1 | - | 1 - 2 | 137 | 67 | |
BT020027.1 | - | 1 - 2 | 37 | 67 | |
BT020028.1 | - | 1 - 2 | 37 | 67 | |
BU944638.1 | - | 1 - 2 | 281 | 67 | |
CR407659.1 | - | 1 - 2 | 37 | 67 | |
U57091.1 | - | 1 - 2 | 82 | 67 | |
U59877.1 | - | 1 - 2 | 97 | 67 |