Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB31
Entrez Gene ID
11031
Gene Name
RAB31, member RAS oncogene family
Gene Aliases
Rab22B
Chromosome Location
Chr.18: 9707971 - 9862556 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006868.3 NP_006859.2 2 - 3 292 71
XM_017025528.1 XP_016881017.1 3 - 4 674 71
XM_017025529.1 XP_016881018.1 3 - 4 662 71
GenBank mRNA AF183421.1 - 2 - 3 245 71
AF234995.1 - 2 - 3 291 71
AF498957.1 - 2 - 3 114 71
AK223505.1 - 2 - 3 156 71
AK314809.1 - 2 - 3 250 71
BC001148.1 - 2 - 3 214 71
BT020027.1 - 2 - 3 114 71
BT020028.1 - 2 - 3 114 71
BU944638.1 - 2 - 3 358 71
CR407659.1 - 2 - 3 114 71
U57091.1 - 2 - 3 159 71
U59877.1 - 2 - 3 174 71

View Full Product Details