The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006579.2 | NP_006570.1 | 1 - 2 | 138 | 116 |
GenBank mRNA | AU128761.1 | - | 1 - 2 | 137 | 114 |
BC001549.1 | - | 1 - 2 | 69 | 115 | |
BC001572.1 | - | 1 - 2 | 69 | 117 | |
CA488777.1 | - | 1 - 2 | 118 | 116 |