The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145039.3 | NP_659476.2 | 1 - 1 | 1355 | 85 |
GenBank mRNA | AK315352.1 | - | 1 - 1 | 988 | 85 |
CD518450.1 | - | 1 - 1 | 393 | 85 |