Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POU2F2
Entrez Gene ID
5452
Gene Name
POU class 2 homeobox 2
Gene Aliases
OCT2, OTF2, Oct-2
Chromosome Location
Chr.19: 42086110 - 42197931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011527040.2 XP_011525342.1 4 - 5 266 77
XM_011527041.2 XP_011525343.1 4 - 5 266 77
XM_011527042.2 XP_011525344.1 4 - 5 266 77
XM_011527043.2 XP_011525345.2 4 - 5 266 77
XM_017026884.1 XP_016882373.1 4 - 5 267 77
XM_017026885.1 XP_016882374.1 4 - 5 270 77
XM_017026886.1 XP_016882375.1 5 - 6 1769 77
XM_017026887.1 XP_016882376.1 5 - 6 239 77
XM_017026888.1 XP_016882377.1 5 - 6 719 77
XM_017026889.1 XP_016882378.1 4 - 5 261 77
XM_017026890.1 XP_016882379.1 4 - 5 342 77
XM_017026891.1 XP_016882380.1 4 - 5 267 77
XM_017026893.1 XP_016882382.1 4 - 5 266 77
XM_017026896.1 XP_016882385.1 4 - 5 267 77

View Full Product Details