The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318195.1 | NP_001305124.1 | 1 - 2 | 196 | 62 |
NM_014222.2 | NP_055037.1 | 1 - 2 | 196 | 62 | |
GenBank mRNA | AF044953.1 | - | 1 - 2 | 121 | 62 |
AK301520.1 | - | 1 - 2 | 164 | 62 | |
AK314135.1 | - | 1 - 2 | 156 | 62 | |
BC001016.2 | - | 1 - 2 | 100 | 62 | |
BU602971.1 | - | 1 - 2 | 198 | 62 |