The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002737.2 | NP_002728.1 | 5 - 6 | 575 | 78 |
XM_017024836.1 | XP_016880325.1 | 5 - 6 | 743 | 78 | |
XM_017024837.1 | XP_016880326.1 | 4 - 5 | 406 | 78 | |
XM_017024838.1 | XP_016880327.1 | 5 - 6 | 502 | 78 | |
XM_017024839.1 | XP_016880328.1 | 4 - 5 | 1441 | 78 | |
XM_017024840.1 | XP_016880329.1 | 5 - 6 | 484 | 78 | |
XM_017024841.1 | XP_016880330.1 | 5 - 6 | 743 | 78 | |
GenBank mRNA | AB209475.1 | - | 6 - 7 | 584 | 78 |
AB451258.1 | - | 5 - 6 | 531 | 78 | |
AB451383.1 | - | 5 - 6 | 531 | 78 | |
AY633609.1 | - | 5 - 6 | 78 | ||
BC053321.1 | - | 5 - 6 | 747 | 78 | |
BC109273.1 | - | 5 - 6 | 557 | 78 | |
BC109274.1 | - | 5 - 6 | 557 | 78 | |
LC151409.1 | - | 5 - 6 | 531 | 78 | |
M22199.1 | - | 5 - 6 | 490 | 78 | |
X52479.1 | - | 5 - 6 | 558 | 78 |