The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002737.2 | NP_002728.1 | 9 - 10 | 1098 | 82 |
XM_017024836.1 | XP_016880325.1 | 9 - 10 | 1266 | 82 | |
XM_017024837.1 | XP_016880326.1 | 8 - 9 | 929 | 82 | |
XM_017024838.1 | XP_016880327.1 | 9 - 10 | 1025 | 82 | |
XM_017024839.1 | XP_016880328.1 | 8 - 9 | 1964 | 82 | |
XM_017024840.1 | XP_016880329.1 | 9 - 10 | 1007 | 82 | |
GenBank mRNA | AB209475.1 | - | 10 - 11 | 1107 | 82 |
AB451258.1 | - | 9 - 10 | 1054 | 82 | |
AB451383.1 | - | 9 - 10 | 1054 | 82 | |
AY633609.1 | - | 9 - 10 | 1059 | 82 | |
BC109273.1 | - | 9 - 10 | 1080 | 82 | |
BC109274.1 | - | 9 - 10 | 1080 | 82 | |
M22199.1 | - | 9 - 10 | 1013 | 82 | |
X52479.1 | - | 9 - 10 | 1081 | 82 |