The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_078476.3 | NP_510961.1 | 7 - 8 | 1251 | 86 |
GenBank mRNA | BC016661.1 | - | 7 - 8 | 1194 | 86 |