The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205247.1 | NP_001192176.1 | 11 - 12 | 1070 | 62 |
NM_001205248.1 | NP_001192177.1 | 11 - 12 | 1162 | 62 | |
NM_015256.3 | NP_056071.2 | 11 - 12 | 1175 | 62 | |
GenBank mRNA | AF102845.1 | - | 10 - 11 | 952 | 62 |
AF129166.1 | - | 11 - 12 | 1105 | 62 | |
AK294291.1 | - | 11 - 12 | 1045 | 62 | |
AK314452.1 | - | 11 - 12 | 1077 | 62 |