The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079855.1 | NP_001073324.1 | 8 - 9 | 1321 | 71 |
NM_001184702.1 | NP_001171631.1 | 8 - 9 | 1321 | 71 | |
NM_003918.2 | NP_003909.2 | 9 - 10 | 1414 | 71 | |
XM_011545600.2 | XP_011543902.1 | 8 - 9 | 1252 | 71 | |
XM_017029927.1 | XP_016885416.1 | 9 - 10 | 1364 | 71 | |
XM_017029928.1 | XP_016885417.1 | 9 - 10 | 1339 | 71 | |
XM_017029929.1 | XP_016885418.1 | 9 - 10 | 1524 | 71 | |
GenBank mRNA | AK292496.1 | - | 8 - 9 | 1229 | 71 |
BC023152.1 | - | 8 - 9 | 1188 | 71 | |
U94358.1 | - | 5 - 6 | 810 | 71 | |
U94361.1 | - | 4 - 5 | 587 | 71 | |
U94362.1 | - | 9 - 10 | 1415 | 71 | |
U94363.1 | - | 8 - 9 | 1195 | 71 |