Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
Chromosome Location
Chr.5: 140841322 - 141012344 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018911.2 NP_061734.1 1 - 2 2395 82
GenBank mRNA AF152316.1 - 1 - 2 2395 82

View Full Product Details