The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033103.4 | NP_149094.3 | 10 - 11 | 1287 | 66 |
GenBank mRNA | AF423421.1 | - | 10 - 11 | 1221 | 66 |
AJ347750.1 | - | 10 - 11 | 1221 | 66 | |
AK098246.1 | - | 10 - 11 | 1257 | 66 | |
AK126506.1 | - | 7 - 8 | 1555 | 66 | |
AK300775.1 | - | 9 - 10 | 1181 | 66 | |
AK315105.1 | - | 10 - 11 | 1257 | 66 | |
AL831950.1 | - | 9 - 10 | 1135 | 66 | |
BC036447.2 | - | 10 - 11 | 1257 | 66 |