The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030928.3 | NP_112190.2 | 5 - 6 | 886 | 142 |
GenBank mRNA | AB053172.1 | - | 5 - 6 | 886 | 142 |
AF321125.1 | - | 5 - 6 | 857 | 142 | |
BC000137.2 | - | 5 - 6 | 848 | 142 | |
BC008676.1 | - | 5 - 6 | 724 | 142 | |
BC008860.2 | - | 5 - 6 | 779 | 142 | |
BC009410.1 | - | 5 - 6 | 842 | 142 | |
BC014202.2 | - | 1 - 2 | 147 | 142 | |
BC021126.1 | - | 3 - 4 | 418 | 142 | |
BC049205.1 | - | 3 - 4 | 490 | 142 |