The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000243.2 | NP_000234.1 | 1 - 2 | 314 | 132 |
XM_017023236.1 | XP_016878725.1 | 1 - 2 | 455 | 132 | |
GenBank mRNA | AF018080.1 | - | 1 - 2 | 315 | 132 |
AK298660.1 | - | 1 - 2 | 308 | 132 | |
BC101511.1 | - | 1 - 2 | 299 | 132 | |
BC101537.1 | - | 1 - 2 | 299 | 132 | |
FJ785721.1 | - | 1 - 2 | 274 | 132 | |
Y14441.1 | - | 1 - 2 | 303 | 132 |