The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000243.2 | NP_000234.1 | 4 - 5 | 1391 | 107 |
NM_001198536.1 | NP_001185465.1 | 3 - 4 | 758 | 107 | |
XM_017023236.1 | XP_016878725.1 | 4 - 5 | 1529 | 107 | |
GenBank mRNA | AF018080.1 | - | 4 - 5 | 1392 | 107 |
AK298660.1 | - | 4 - 5 | 1385 | 107 | |
BC101511.1 | - | 4 - 5 | 1376 | 107 | |
BC101537.1 | - | 4 - 5 | 1376 | 107 | |
FJ785722.1 | - | 3 - 4 | 718 | 107 | |
Y14441.1 | - | 4 - 5 | 1380 | 107 |