Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM57B
Entrez Gene ID
83723
Gene Name
family with sequence similarity 57 member B
Gene Aliases
FP1188
Chromosome Location
Chr.16: 30024423 - 30053026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005255613.3 XP_005255670.1 1 - 2 260 77
XM_005255614.3 XP_005255671.1 1 - 2 260 77
XM_017023752.1 XP_016879241.1 1 - 2 260 77

View Full Product Details