Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM57B
Entrez Gene ID
83723
Gene Name
family with sequence similarity 57 member B
Gene Aliases
FP1188
Chromosome Location
Chr.16: 30024423 - 30053026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005255613.3 XP_005255670.1 2 - 3 320 116
XM_005255614.3 XP_005255671.1 2 - 3 320 116
XM_005255615.2 XP_005255672.1 2 - 3 517 116
XM_017023750.1 XP_016879239.1 2 - 3 517 116
XM_017023751.1 XP_016879240.1 3 - 4 413 116
XM_017023752.1 XP_016879241.1 2 - 3 320 116

View Full Product Details