Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLE4
Entrez Gene ID
7091
Gene Name
transducin like enhancer of split 4
Gene Aliases
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
Chromosome Location
Chr.9: 79571773 - 79726882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282748.1 NP_001269677.1 10 - 11 1805 112
NM_001282749.1 NP_001269678.1 9 - 10 1730 112
NM_001282760.1 NP_001269689.1 11 - 12 1900 112
NM_007005.4 NP_008936.2 10 - 11 1805 112
NR_104239.1 11 - 12 1938 112
XM_005252177.1 XP_005252234.1 10 - 11 981 112
XM_006717264.1 XP_006717327.1 12 - 13 1104 112
XM_006717268.1 XP_006717331.1 12 - 13 1104 112
XM_011518952.1 XP_011517254.1 12 - 13 1104 112
XM_011518953.1 XP_011517255.1 12 - 13 981 112
XM_011518954.1 XP_011517256.1 11 - 12 1059 112
XM_011518955.1 XP_011517257.1 11 - 12 1041 112
XM_011518956.1 XP_011517258.1 11 - 12 1020 112
XM_011518957.1 XP_011517259.1 11 - 12 1017 112
XM_011518958.1 XP_011517260.1 12 - 13 1104 112
XM_011518959.1 XP_011517261.1 10 - 11 981 112
XM_011518960.1 XP_011517262.1 10 - 11 978 112
XM_011518961.1 XP_011517263.1 9 - 10 918 112
XM_011518962.1 XP_011517264.1 9 - 10 915 112
XM_011518963.1 XP_011517265.1 9 - 10 906 112
XM_011518964.1 XP_011517266.1 9 - 10 903 112
XM_011518965.1 XP_011517267.1 10 - 11 981 112
XM_011518966.1 XP_011517268.1 10 - 11 978 112
XM_011518968.1 XP_011517270.1 8 - 9 843 112
XM_011518970.2 XP_011517272.1 12 - 13 1104 112
XM_017015067.1 XP_016870556.1 10 - 11 909 112
XM_017015068.1 XP_016870557.1 9 - 10 936 112
XM_017015069.1 XP_016870558.1 9 - 10 764 112
XM_017015070.1 XP_016870559.1 9 - 10 841 112
XM_017015071.1 XP_016870560.1 11 - 12 1017 112
XM_017015072.1 XP_016870561.1 10 - 11 938 112
XM_017015073.1 XP_016870562.1 10 - 11 940 112
XM_017015075.1 XP_016870564.1 9 - 10 933 112
XM_017015076.1 XP_016870565.1 9 - 10 880 112
XM_017015077.1 XP_016870566.1 9 - 10 918 112
XM_017015078.1 XP_016870567.1 9 - 10 915 112
XM_017015079.1 XP_016870568.1 9 - 10 843 112
XM_017015080.1 XP_016870569.1 9 - 10 906 112
XM_017015082.1 XP_016870571.1 9 - 10 798 112
GenBank mRNA AB033087.1 - 10 - 11 866 112
AK057236.1 - 11 - 12 1086 112
AK296342.1 - 9 - 10 759 112
AK302709.1 - 10 - 11 840 112
AL162059.1 - 8 - 9 640 112
BC036369.1 - 11 - 12 1100 112
BC059405.1 - 10 - 11 963 112

View Full Product Details