Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TLE4
Entrez Gene ID
7091
Gene Name
transducin like enhancer of split 4
Gene Aliases
BCE-1, BCE1, E(spI), ESG, ESG4, GRG4, Grg-4
Chromosome Location
Chr.9: 79571773 - 79726882 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282748.1 NP_001269677.1 5 - 6 1336 61
NM_001282753.1 NP_001269682.1 5 - 6 1336 61
NM_001282760.1 NP_001269689.1 6 - 7 1431 61
NM_007005.4 NP_008936.2 5 - 6 1336 61
NR_104239.1 5 - 6 1336 61
XM_005252177.1 XP_005252234.1 5 - 6 509 61
XM_006717264.1 XP_006717327.1 5 - 6 509 61
XM_006717268.1 XP_006717331.1 5 - 6 509 61
XM_011518952.1 XP_011517254.1 5 - 6 509 61
XM_011518953.1 XP_011517255.1 5 - 6 386 61
XM_011518954.1 XP_011517256.1 4 - 5 464 61
XM_011518956.1 XP_011517258.1 5 - 6 509 61
XM_011518957.1 XP_011517259.1 5 - 6 509 61
XM_011518958.1 XP_011517260.1 5 - 6 509 61
XM_011518959.1 XP_011517261.1 5 - 6 509 61
XM_011518960.1 XP_011517262.1 5 - 6 509 61
XM_011518965.1 XP_011517267.1 5 - 6 509 61
XM_011518966.1 XP_011517268.1 5 - 6 509 61
XM_011518967.1 XP_011517269.1 5 - 6 509 61
XM_011518969.1 XP_011517271.1 5 - 6 509 61
XM_011518970.2 XP_011517272.1 5 - 6 509 61
XM_017015067.1 XP_016870556.1 5 - 6 437 61
XM_017015068.1 XP_016870557.1 4 - 5 464 61
XM_017015071.1 XP_016870560.1 5 - 6 509 61
XM_017015072.1 XP_016870561.1 5 - 6 466 61
XM_017015073.1 XP_016870562.1 5 - 6 471 61
XM_017015074.1 XP_016870563.1 5 - 6 509 61
XM_017015075.1 XP_016870564.1 4 - 5 464 61
XM_017015076.1 XP_016870565.1 4 - 5 411 61
XM_017015081.1 XP_016870570.1 5 - 6 509 61
XM_017015083.1 XP_016870572.1 5 - 6 509 61
GenBank mRNA AB033087.1 - 5 - 6 397 61
AK057236.1 - 5 - 6 484 61
AK302709.1 - 5 - 6 371 61
AL162059.1 - 3 - 4 168 61
BC036369.1 - 6 - 7 631 61
BC045650.1 - 5 - 6 1336 61
BC059405.1 - 5 - 6 494 61

View Full Product Details