Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA4D
Entrez Gene ID
10507
Gene Name
semaphorin 4D
Gene Aliases
C9orf164, CD100, M-sema-G, SEMAJ, coll-4
Chromosome Location
Chr.9: 89360791 - 89498065 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142287.1 NP_001135759.1 13 - 14 1678 101
NM_006378.3 NP_006369.3 13 - 14 1678 101
XM_005251654.3 XP_005251711.1 11 - 12 1512 101
XM_011518123.2 XP_011516425.1 15 - 16 1762 101
XM_011518124.2 XP_011516426.1 15 - 16 1721 101
XM_011518125.1 XP_011516427.1 13 - 14 1649 101
XM_011518127.2 XP_011516429.1 14 - 15 1677 101
XM_011518128.2 XP_011516430.1 13 - 14 1602 101
XM_011518129.1 XP_011516431.1 12 - 13 1531 101
XM_011518130.2 XP_011516432.1 12 - 13 1766 101
XM_011518131.2 XP_011516433.1 11 - 12 1704 101
XM_011518133.2 XP_011516435.1 12 - 13 1628 101
XM_011518134.2 XP_011516436.1 10 - 11 1569 101
XM_017014193.1 XP_016869682.1 14 - 15 3372 101
XM_017014194.1 XP_016869683.1 14 - 15 1646 101
XM_017014195.1 XP_016869684.1 13 - 14 1583 101
XM_017014196.1 XP_016869685.1 13 - 14 1920 101
XM_017014197.1 XP_016869686.1 12 - 13 1804 101
XM_017014198.1 XP_016869687.1 11 - 12 1741 101
XM_017014199.1 XP_016869688.1 15 - 16 1721 101
GenBank mRNA AB210030.1 - 11 - 12 1883 101
AK293749.1 - 13 - 14 1788 101
AK316107.1 - 13 - 14 1872 101
BC054500.1 - 13 - 14 1678 101
BX648216.1 - 11 - 12 1470 101
U60800.1 - 9 - 10 1193 101

View Full Product Details