Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA4D
Entrez Gene ID
10507
Gene Name
semaphorin 4D
Gene Aliases
C9orf164, CD100, M-sema-G, SEMAJ, coll-4
Chromosome Location
Chr.9: 89360791 - 89498065 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142287.1 NP_001135759.1 9 - 10 1081 93
NM_006378.3 NP_006369.3 9 - 10 1081 93
XM_005251654.3 XP_005251711.1 7 - 8 915 93
XM_011518123.2 XP_011516425.1 11 - 12 1165 93
XM_011518124.2 XP_011516426.1 11 - 12 1124 93
XM_011518125.1 XP_011516427.1 9 - 10 1052 93
XM_011518127.2 XP_011516429.1 10 - 11 1080 93
XM_011518128.2 XP_011516430.1 9 - 10 1005 93
XM_011518129.1 XP_011516431.1 8 - 9 934 93
XM_011518130.2 XP_011516432.1 8 - 9 1169 93
XM_011518131.2 XP_011516433.1 7 - 8 1107 93
XM_011518133.2 XP_011516435.1 8 - 9 1031 93
XM_011518134.2 XP_011516436.1 6 - 7 972 93
XM_017014193.1 XP_016869682.1 10 - 11 2775 93
XM_017014194.1 XP_016869683.1 10 - 11 1049 93
XM_017014195.1 XP_016869684.1 9 - 10 986 93
XM_017014196.1 XP_016869685.1 9 - 10 1323 93
XM_017014197.1 XP_016869686.1 8 - 9 1207 93
XM_017014198.1 XP_016869687.1 7 - 8 1144 93
XM_017014199.1 XP_016869688.1 11 - 12 1124 93
GenBank mRNA AB210030.1 - 7 - 8 1286 93
AK293749.1 - 8 - 10 1191 93
BC054500.1 - 9 - 10 1081 93
BX648216.1 - 7 - 8 873 93
U60800.1 - 5 - 6 596 93

View Full Product Details