Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEMA4D
Entrez Gene ID
10507
Gene Name
semaphorin 4D
Gene Aliases
C9orf164, CD100, M-sema-G, SEMAJ, coll-4
Chromosome Location
Chr.9: 89360791 - 89498065 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006378.3 NP_006369.3 17 - 18 2234 60
XM_005251654.3 XP_005251711.1 15 - 16 2068 60
XM_011518123.2 XP_011516425.1 19 - 20 2318 60
XM_011518124.2 XP_011516426.1 19 - 20 2277 60
XM_011518125.1 XP_011516427.1 17 - 18 2205 60
XM_011518127.2 XP_011516429.1 18 - 19 2233 60
XM_011518128.2 XP_011516430.1 17 - 18 2158 60
XM_011518129.1 XP_011516431.1 16 - 17 2087 60
XM_011518130.2 XP_011516432.1 16 - 17 2322 60
XM_011518131.2 XP_011516433.1 15 - 16 2260 60
XM_011518133.2 XP_011516435.1 16 - 17 2184 60
XM_011518134.2 XP_011516436.1 14 - 15 2125 60
XM_017014193.1 XP_016869682.1 18 - 19 3928 60
XM_017014194.1 XP_016869683.1 18 - 19 2202 60
XM_017014195.1 XP_016869684.1 17 - 18 2139 60
XM_017014196.1 XP_016869685.1 17 - 18 2476 60
XM_017014197.1 XP_016869686.1 16 - 17 2360 60
XM_017014198.1 XP_016869687.1 15 - 16 2297 60
GenBank mRNA AB210030.1 - 15 - 16 2439 60
BC054500.1 - 17 - 18 2234 60
BX648216.1 - 15 - 16 2026 60
U60800.1 - 13 - 14 1749 60

View Full Product Details