Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
Chromosome Location
Chr.17: 67825517 - 67984378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004459.6 NP_004450.3 19 - 20 6586 68
NM_182641.3 NP_872579.2 17 - 18 6208 68
XM_005257150.3 XP_005257207.1 20 - 21 6895 68
XM_005257151.3 XP_005257208.1 20 - 21 6892 68
XM_005257152.2 XP_005257209.1 20 - 21 6895 68
XM_005257153.3 XP_005257210.1 19 - 20 6733 68
XM_005257154.3 XP_005257211.1 20 - 21 6895 68
XM_005257155.3 XP_005257212.1 19 - 20 6706 68
XM_005257156.3 XP_005257213.1 19 - 20 6706 68
XM_005257157.3 XP_005257214.1 19 - 20 6703 68
XM_005257158.3 XP_005257215.1 18 - 19 6517 68
XM_005257159.2 XP_005257216.1 20 - 21 6895 68
XM_005257160.2 XP_005257217.1 19 - 20 6733 68
XM_005257161.3 XP_005257218.1 20 - 21 6895 68
XM_011524520.2 XP_011522822.1 20 - 21 6892 68
XM_011524521.2 XP_011522823.1 19 - 20 6703 68
XM_011524522.2 XP_011522824.1 18 - 19 6514 68
XM_011524523.2 XP_011522825.1 17 - 18 6352 68
XM_011524524.2 XP_011522826.1 12 - 13 3856 68
XM_011524525.2 XP_011522827.1 17 - 18 6352 68
XM_011524526.2 XP_011522828.1 17 - 18 6352 68
XM_017024353.1 XP_016879842.1 20 - 21 6895 68
XM_017024354.1 XP_016879843.1 14 - 15 5624 68
GenBank mRNA AB032251.1 - 17 - 18 6201 68
AK022570.1 - 5 - 6 686 68
AK025776.1 - 7 - 8 2031 68
AY282495.1 - 19 - 20 6108 68
BQ716216.1 - 4 - 5 558 68

View Full Product Details