Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
Chromosome Location
Chr.17: 67825517 - 67984378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004459.6 NP_004450.3 27 - 28 8458 81
NM_182641.3 NP_872579.2 25 - 26 8509 81
XM_005257150.3 XP_005257207.1 29 - 30 9370 81
XM_005257151.3 XP_005257208.1 29 - 30 9367 81
XM_005257152.2 XP_005257209.1 29 - 30 9370 81
XM_005257153.3 XP_005257210.1 28 - 29 9208 81
XM_005257154.3 XP_005257211.1 28 - 29 9196 81
XM_005257155.3 XP_005257212.1 28 - 29 9181 81
XM_005257156.3 XP_005257213.1 28 - 29 9181 81
XM_005257157.3 XP_005257214.1 28 - 29 9178 81
XM_005257158.3 XP_005257215.1 27 - 28 8992 81
XM_005257159.2 XP_005257216.1 29 - 30 8941 81
XM_005257160.2 XP_005257217.1 28 - 29 8779 81
XM_005257161.3 XP_005257218.1 28 - 29 8767 81
XM_011524520.2 XP_011522822.1 29 - 30 9367 81
XM_011524521.2 XP_011522823.1 28 - 29 9178 81
XM_011524522.2 XP_011522824.1 27 - 28 8989 81
XM_011524523.2 XP_011522825.1 26 - 27 8827 81
XM_011524524.2 XP_011522826.1 21 - 22 6331 81
XM_011524525.2 XP_011522827.1 25 - 26 8653 81
XM_011524526.2 XP_011522828.1 25 - 26 8224 81
XM_017024353.1 XP_016879842.1 28 - 29 9196 81
XM_017024354.1 XP_016879843.1 23 - 24 8099 81
GenBank mRNA AB032251.1 - 25 - 26 8502 81
AK296256.1 - 6 - 7 1935 81
AL833375.1 - 7 - 8 1648 81
AY282495.1 - 27 - 28 7980 81
BC050566.1 - 3 - 4 410 81
BC067234.1 - 1 - 2 201 81

View Full Product Details