Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BPTF
Entrez Gene ID
2186
Gene Name
bromodomain PHD finger transcription factor
Gene Aliases
FAC1, FALZ, NURF301
Chromosome Location
Chr.17: 67825517 - 67984378 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004459.6 NP_004450.3 28 - 29 8542 115
NM_182641.3 NP_872579.2 26 - 27 8593 115
XM_005257150.3 XP_005257207.1 30 - 31 9454 115
XM_005257151.3 XP_005257208.1 30 - 31 9451 115
XM_005257152.2 XP_005257209.1 30 - 31 9454 115
XM_005257153.3 XP_005257210.1 29 - 30 9292 115
XM_005257154.3 XP_005257211.1 29 - 30 9280 115
XM_005257155.3 XP_005257212.1 29 - 30 9265 115
XM_005257156.3 XP_005257213.1 29 - 30 9265 115
XM_005257157.3 XP_005257214.1 29 - 30 9262 115
XM_005257158.3 XP_005257215.1 28 - 29 9076 115
XM_005257159.2 XP_005257216.1 30 - 31 9025 115
XM_005257160.2 XP_005257217.1 29 - 30 8863 115
XM_005257161.3 XP_005257218.1 29 - 30 8851 115
XM_011524520.2 XP_011522822.1 30 - 31 9451 115
XM_011524521.2 XP_011522823.1 29 - 30 9262 115
XM_011524522.2 XP_011522824.1 28 - 29 9073 115
XM_011524523.2 XP_011522825.1 27 - 28 8911 115
XM_011524524.2 XP_011522826.1 22 - 23 6415 115
XM_011524525.2 XP_011522827.1 26 - 27 8737 115
XM_011524526.2 XP_011522828.1 26 - 27 8308 115
XM_017024353.1 XP_016879842.1 29 - 30 9280 115
XM_017024354.1 XP_016879843.1 24 - 25 8183 115
GenBank mRNA AB032251.1 - 26 - 27 8586 115
AK022081.1 - 1 - 2 1629 115
AK296256.1 - 7 - 8 2019 115
AL833375.1 - 8 - 9 1732 115
AY282495.1 - 28 - 29 8064 115
BC050566.1 - 4 - 5 494 115
BC067234.1 - 2 - 3 285 115

View Full Product Details