Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIOX
Entrez Gene ID
55586
Gene Name
myo-inositol oxygenase
Gene Aliases
ALDRL6
Chromosome Location
Chr.22: 50483988 - 50490321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017584.5 NP_060054.4 4 - 5 455 110
XM_005261925.3 XP_005261982.1 3 - 4 322 110
GenBank mRNA AF197129.1 - 3 - 4 341 110
AY064416.1 - 4 - 5 368 110
AY738258.1 - 3 - 4 341 110
BC064395.1 - 4 - 5 363 110
BC073848.1 - 4 - 5 375 110
CR456478.1 - 4 - 5 400 110

View Full Product Details