The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022785.3 | NP_073622.2 | 31 - 32 | 4701 | 92 |
NM_198856.2 | NP_942153.1 | 29 - 30 | 4343 | 92 | |
XM_011530320.2 | XP_011528622.1 | 32 - 33 | 4736 | 92 | |
XM_017028911.1 | XP_016884400.1 | 32 - 33 | 4740 | 92 | |
GenBank mRNA | AA778573.1 | - | 1 - 2 | 92 | |
AB073862.1 | - | 11 - 12 | 1895 | 92 | |
AK027241.1 | - | 11 - 12 | 1899 | 92 | |
AK225596.1 | - | 11 - 12 | 1900 | 92 | |
AK225605.1 | - | 11 - 12 | 1900 | 92 | |
AK292411.1 | - | 31 - 32 | 4641 | 92 | |
AK292498.1 | - | 29 - 30 | 4320 | 92 | |
AL355841.1 | - | 6 - 7 | 1024 | 92 | |
AW139921.1 | - | 1 - 2 | 92 | ||
BC039315.1 | - | 29 - 30 | 4196 | 92 |