The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145212.3 | NP_660213.1 | 1 - 2 | 169 | 81 |
NR_028356.1 | 1 - 2 | 169 | 81 | ||
GenBank mRNA | AK025547.1 | - | 1 - 2 | 96 | 81 |
AK225894.1 | - | 1 - 2 | 65 | 81 | |
BC000217.1 | - | 1 - 2 | 59 | 81 | |
BC022391.1 | - | 1 - 2 | 133 | 81 | |
BG532984.1 | - | 1 - 2 | 51 | 81 | |
BU154006.1 | - | - | 42 | 81 | |
BU533068.1 | - | 1 - 2 | 99 | 81 | |
DA310406.1 | - | 1 - 2 | 82 |