Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
POLR3E
Entrez Gene ID
55718
Gene Name
RNA polymerase III subunit E
Gene Aliases
RPC5, SIN
Chromosome Location
Chr.16: 22297366 - 22335103 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001258033.1 NP_001244962.1 6 - 7 569 63
NM_001258034.1 NP_001244963.1 5 - 6 461 63
NM_001258035.1 NP_001244964.1 5 - 6 495 63
NM_001258036.1 NP_001244965.1 6 - 7 569 63
NM_018119.3 NP_060589.1 6 - 7 569 63
NR_047581.1 6 - 7 519 63
XM_006721063.2 XP_006721126.1 6 - 7 490 63
XM_011545892.1 XP_011544194.1 6 - 7 578 63
XM_011545893.2 XP_011544195.1 6 - 7 578 63
GenBank mRNA AB040885.1 - 6 - 7 489 63
AK001371.1 - 6 - 7 510 63
AK023160.1 - 6 - 7 524 63
AK023231.1 - 5 - 6 447 63
AK026645.1 - 6 - 7 535 63
AK222671.1 - 6 - 7 510 63
AK222789.1 - 6 - 7 516 63
AK293300.1 - 5 - 6 340 63
AK296813.1 - 5 - 6 409 63
AY092085.1 - 5 - 6 368 63
BC000285.1 - 6 - 7 511 63

View Full Product Details