The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258033.1 | NP_001244962.1 | 20 - 21 | 2206 | 131 |
NM_001258034.1 | NP_001244963.1 | 19 - 20 | 2161 | 131 | |
NM_001258035.1 | NP_001244964.1 | 19 - 20 | 2195 | 131 | |
NM_018119.3 | NP_060589.1 | 20 - 21 | 2269 | 131 | |
NR_047581.1 | 20 - 21 | 2219 | 131 | ||
XM_006721063.2 | XP_006721126.1 | 20 - 21 | 2190 | 131 | |
XM_011545892.1 | XP_011544194.1 | 20 - 21 | 2275 | 131 | |
XM_017023443.1 | XP_016878932.1 | 20 - 21 | 2273 | 131 | |
GenBank mRNA | AK001371.1 | - | 20 - 21 | 2210 | 131 |
AK023231.1 | - | 19 - 20 | 2147 | 131 | |
AK026645.1 | - | 20 - 21 | 2235 | 131 | |
AK222789.1 | - | 20 - 21 | 2216 | 131 | |
AK293300.1 | - | 19 - 20 | 2040 | 131 | |
AK300735.1 | - | 20 - 21 | 2170 | 131 |