Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
Chromosome Location
Chr.18: 3496032 - 4455307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242764.1 NP_001229693.1 1 - 2 95 78
NM_001308390.1 NP_001295319.1 1 - 2 147 78
XM_017026085.1 XP_016881574.1 1 - 2 357 78
GenBank mRNA AK299880.1 - 1 - 2 95 78

View Full Product Details