Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLGAP1
Entrez Gene ID
9229
Gene Name
DLG associated protein 1
Gene Aliases
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1
Chromosome Location
Chr.18: 3496032 - 4455307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242763.1 NP_001229692.1 4 - 5 790 67
NM_001242766.1 NP_001229695.1 4 - 5 763 67
NM_001308390.1 NP_001295319.1 4 - 5 788 67
XM_005258171.2 XP_005258228.1 7 - 8 2161 67
XM_005258172.1 XP_005258229.1 7 - 8 2156 67
XM_005258173.3 XP_005258230.2 5 - 6 952 67
XM_005258174.4 XP_005258231.1 4 - 5 1009 67
XM_006722367.3 XP_006722430.1 5 - 6 1120 67
GenBank mRNA AK294747.1 - 4 - 5 763 67
AK294777.1 - 4 - 5 713 67

View Full Product Details