Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYN2
Entrez Gene ID
6854
Gene Name
synapsin II
Gene Aliases
SYNII
Chromosome Location
Chr.3: 12004360 - 12192032 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133625.4 NP_598328.1 12 - 13 1804 79
XM_006713312.3 XP_006713375.1 9 - 10 2006 79
XM_006713313.2 XP_006713376.1 9 - 10 1144 79
XM_017007087.1 XP_016862576.1 8 - 9 1216 79
GenBank mRNA AF077671.1 - 12 - 13 1633 79
AK091286.1 - 9 - 10 1231 79
AK094464.1 - 13 - 14 1572 79
BC051307.1 - 12 - 13 1762 79

View Full Product Details