The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199867.1 | NP_001186796.1 | 5 - 6 | 753 | 60 |
NM_031417.3 | NP_113605.2 | 5 - 6 | 753 | 60 | |
XM_006723307.3 | XP_006723370.1 | 4 - 5 | 444 | 60 | |
GenBank mRNA | AB049127.1 | - | 5 - 6 | 719 | 60 |
AB058763.1 | - | 5 - 6 | 427 | 60 | |
AB088047.1 | - | 5 - 6 | 719 | 60 | |
AK074578.1 | - | 1 - 2 | 64 | 60 | |
AK075272.1 | - | 5 - 6 | 458 | 60 | |
AY057448.1 | - | 5 - 6 | 434 | 60 | |
AY120867.1 | - | 5 - 6 | 422 | 60 |