Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDFY4
Entrez Gene ID
57705
Gene Name
WDFY family member 4
Gene Aliases
C10orf64
Chromosome Location
Chr.10: 48684815 - 48982956 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020945.1 NP_065996.1 52 - 53 8207 83
XM_005270004.3 XP_005270061.1 52 - 53 8322 83
XM_011539986.2 XP_011538288.2 52 - 53 8325 83
XM_011539987.2 XP_011538289.2 52 - 53 8328 83
XM_011539988.2 XP_011538290.1 53 - 54 8406 83
XM_017016463.1 XP_016871952.1 53 - 54 8249 83
XM_017016464.1 XP_016871953.1 34 - 35 4636 83
GenBank mRNA AB046827.1 - 20 - 21 2390 83
AK024502.1 - 3 - 4 374 83
AK074085.1 - 33 - 34 4265 83
AK074443.1 - 15 - 16 1610 83
AK301736.1 - 6 - 7 663 83
AK310374.1 - 6 - 7 707 83
AL833669.1 - 13 - 14 1336 83
BC047574.1 - 13 - 14 1276 83

View Full Product Details