The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322500.1 | NP_001309429.1 | 12 - 13 | 1654 | 100 |
NM_001322501.1 | NP_001309430.1 | 11 - 12 | 1578 | 100 | |
NM_017886.3 | NP_060356.2 | 12 - 13 | 1654 | 100 | |
NR_136342.1 | 11 - 12 | 1584 | 100 | ||
XM_011533874.2 | XP_011532176.1 | 12 - 13 | 2232 | 100 | |
XM_017006711.1 | XP_016862200.1 | 12 - 13 | 2232 | 100 | |
XM_017006712.1 | XP_016862201.1 | 12 - 13 | 2229 | 100 | |
GenBank mRNA | AK000581.1 | - | - | 1293 | 100 |
AK304341.1 | - | 12 - 13 | 1285 | 100 | |
BC014794.1 | - | 12 - 13 | 1273 | 100 |