The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322500.1 | NP_001309429.1 | 18 - 19 | 2239 | 152 |
NM_001322501.1 | NP_001309430.1 | 17 - 18 | 2163 | 152 | |
NM_017886.3 | NP_060356.2 | 18 - 19 | 2239 | 152 | |
NR_136342.1 | 17 - 18 | 2169 | 152 | ||
XM_011533874.2 | XP_011532176.1 | 18 - 19 | 2817 | 152 | |
XM_017006712.1 | XP_016862201.1 | 18 - 19 | 2814 | 152 | |
GenBank mRNA | AK000581.1 | - | - | 1879 | 152 |
AK304341.1 | - | 18 - 19 | 1870 | 152 | |
BX096790.1 | - | 4 - 5 | 255 | 152 |