The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005259010.3 | XP_005259067.1 | 13 - 14 | 1428 | 55 |
XM_011527040.2 | XP_011525342.1 | 13 - 14 | 1542 | 55 | |
XM_017026884.1 | XP_016882373.1 | 14 - 15 | 1840 | 55 | |
XM_017026885.1 | XP_016882374.1 | 14 - 15 | 1843 | 55 | |
XM_017026886.1 | XP_016882375.1 | 15 - 16 | 3342 | 55 | |
XM_017026887.1 | XP_016882376.1 | 15 - 16 | 1812 | 55 | |
XM_017026888.1 | XP_016882377.1 | 15 - 16 | 2292 | 55 | |
XM_017026889.1 | XP_016882378.1 | 14 - 15 | 1834 | 55 | |
XM_017026890.1 | XP_016882379.1 | 14 - 15 | 1915 | 55 | |
XM_017026891.1 | XP_016882380.1 | 14 - 15 | 1792 | 55 | |
XM_017026892.1 | XP_016882381.1 | 14 - 15 | 1774 | 55 | |
XM_017026893.1 | XP_016882382.1 | 13 - 14 | 1494 | 55 | |
XM_017026894.1 | XP_016882383.1 | 13 - 14 | 1476 | 55 | |
XM_017026895.1 | XP_016882384.1 | 13 - 14 | 1474 | 55 |