Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2F2
Entrez Gene ID
5452
Gene Name
POU class 2 homeobox 2
Gene Aliases
OCT2, OTF2, Oct-2
Chromosome Location
Chr.19: 42086110 - 42197931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005259010.3 XP_005259067.1 14 - 15 1511 79
XM_011527040.2 XP_011525342.1 14 - 15 1625 79
XM_017026884.1 XP_016882373.1 15 - 16 1923 79
XM_017026885.1 XP_016882374.1 15 - 16 1926 79
XM_017026886.1 XP_016882375.1 16 - 17 3425 79
XM_017026887.1 XP_016882376.1 16 - 17 1895 79
XM_017026888.1 XP_016882377.1 16 - 17 2375 79
XM_017026889.1 XP_016882378.1 15 - 16 1917 79
XM_017026890.1 XP_016882379.1 15 - 16 1998 79
XM_017026891.1 XP_016882380.1 15 - 16 1875 79
XM_017026892.1 XP_016882381.1 15 - 16 1857 79
XM_017026893.1 XP_016882382.1 14 - 15 1577 79
XM_017026894.1 XP_016882383.1 14 - 15 1559 79
XM_017026895.1 XP_016882384.1 14 - 15 1557 79

View Full Product Details