Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RFT1
Entrez Gene ID
91869
Gene Name
RFT1 homolog
Gene Aliases
CDG1N
Chromosome Location
Chr.3: 53071151 - 53130469 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052859.3 NP_443091.1 13 - 13 3709 155
GenBank mRNA AK098811.1 - 1 - 1 901 155
AL713685.1 - 1 - 1 848 155
BC101247.2 - 1 - 1 774 155
BC101248.2 - 1 - 1 774 155
BC101249.2 - 1 - 1 774 155
BC101250.1 - 1 - 1 774 155
BC113058.1 - 1 - 1 774 155
BC113059.1 - 1 - 1 774 155

View Full Product Details