Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCG5
Entrez Gene ID
6447
Gene Name
secretogranin V
Gene Aliases
7B2, P7B2, SGNE1, SgV
Chromosome Location
Chr.15: 32641669 - 32697097 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144757.1 NP_001138229.1 1 - 2 112 74
NM_003020.3 NP_003011.1 1 - 2 112 74
GenBank mRNA AK314965.1 - 1 - 2 105 74
BC005349.1 - 1 - 2 51 74
BC093053.1 - 1 - 2 71 74
BP377248.1 - 1 - 2 112 74

View Full Product Details