The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012321.4 | NP_036453.1 | 2 - 3 | 219 | 86 |
GenBank mRNA | AF117235.1 | - | 2 - 3 | 182 | 86 |
AF182290.1 | - | 2 - 3 | 96 | 86 | |
AF251218.1 | - | 2 - 3 | 90 | 86 | |
AJ238096.1 | - | 2 - 3 | 71 | 86 | |
BC000387.2 | - | 2 - 3 | 99 | 86 | |
BC003652.2 | - | 2 - 3 | 97 | 86 | |
BC022198.2 | - | 2 - 3 | 97 | 86 | |
BC023665.2 | - | 2 - 3 | 92 | 86 | |
DB493307.1 | - | 2 - 3 | 302 | 86 |