The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017514.4 | NP_059984.3 | 18 - 19 | 3368 | 69 |
XM_005274705.4 | XP_005274762.2 | 18 - 19 | 3308 | 69 | |
XM_005274706.4 | XP_005274763.2 | 18 - 19 | 3040 | 69 | |
XM_006724829.3 | XP_006724892.1 | 18 - 19 | 3272 | 69 | |
XM_011531183.2 | XP_011529485.1 | 17 - 18 | 2732 | 69 | |
GenBank mRNA | BQ718472.1 | - | 5 - 6 | 563 | 69 |
X87852.1 | - | 18 - 19 | 3377 | 69 |